Läs mer
Losartan Carboxylic Acid Acyl-Beta-D-Glucuronide, 90%, TRC
Produktkod. 30482640
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30482640 1 mg
1 options
Produktkod. 30482640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4705151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Losartan Carboxylic Acid Acyl-beta-D-Glucuronide
CAS 1799830-02-6
Molekylformel C28H29ClN6O8
Kvantitet 1 mg
Synonym Losartan Carboxylic Acid Glucuronide
InChI formel InChI=1S/C28H29ClN6O8/c1-2-3-8-18-30-24(29)19(27(41)43-28-22(38)20(36)21(37)23(42-28)26(39)40)35(18)13-14-9-11-15(12-10-14)16-6-4-5-7-17(16)25-31-33-34-32-25/h4-7,9-12,20-23,28,36-38H,2-3,8,13H2,1H3,(H,39,40)(H,31,32,33,34)/t20-,21-,22+,23-,28+/m1/s1
LEDER ClC1=C(C(O[C@H]2[C@@H](O)[C@H](O)[C@@H](O)[C@H](C(O)=O)O2)=O)N(CC(C=C3)=CC=C3C4=C(C5=NNN=N5)C=CC=C4)C(CCCC)=N1
Molekylvikt (g/mol) 613.02