Läs mer
Losartan Carboxylic Acid, TRC
Produktkod. 30008462
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008462 50 mg
30005864 1 mg
30005821 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008462 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47051050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124750-92-1
Molekylformel C22 H21 Cl N6 O2
Molekylvikt (g/mol) 436.89
Synonym 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid, E 3174, EXP 3174, Losartan Acid, 1H-Imidazole-5-carboxylic acid, 2-butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 2-Butyl-5-chloro-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid, Losartan carboxylic acid, Losartan-carboxylic acid
IUPAC-namn 2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
LEDER CCCCc1nc(Cl)c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
Specifikationer
Kemiskt namn eller material Losartan Carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H21ClN6O2/c1-2-3-8-18-24-20(23)19(22(30)31)29(18)13-14-9-11-15(12-10-14)16-6-4-5-7-17(16)21-25-27-28-26-21/h4-7,9-12H,2-3,8,13H2,1H3,(H,30,31)(H,25,26,27,28)
Formel vikt 436.1415
Procent renhet >95%