missing translation for 'onlineSavingsMsg'
Läs mer

Losartan Imidazo[1,5-b]isoquinoline Impurity, TRC

Produktkod. 30471699
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471699 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30471699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47054010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Losartan Impurity G
CAS 165276-38-0
Molekylformel C22H21ClN6
Kvantitet 10 mg
InChI formel InChI=1S/C22H21ClN6/c1-2-3-8-20-24-21(23)19-12-16-11-14(9-10-15(16)13-29(19)20)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-11H,2-3,8,12-13H2,1H3,(H,25,26,27,28)
LEDER CCCCc1nc(Cl)c2Cc3cc(ccc3Cn12)c4ccccc4c5nn[nH]n5
IUPAC-namn 3-butyl-1-chloro-8-[2-(2H-tetrazol-5-yl)phenyl]-5,10-dihydroimidazo[1,5-b]isoquinoline
Molekylvikt (g/mol) 404.9
Formel vikt 404.1516
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.