Läs mer
Losartan, TRC
Produktkod. 30136807
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136807 100 mg
18060910 25 mg
18060420 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136807 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL470470100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 114798-26-4
Molekylformel C22 H23 Cl N6 O
Molekylvikt (g/mol) 422.91
Synonym Losartan, 2-Butyl-4-chloro-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-methanol, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-methanol, 2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl]methyl]-1H-imidazole-5-methanol, DUP 89, Lortaan
IUPAC-namn [2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
LEDER CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
Specifikationer
Kemiskt namn eller material Losartan
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C22H23ClN6O/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,30H,2-3,8,13-14H2,1H3,(H,25,26,27,28)
Formel vikt 422.1622
Procent renhet >95%