Läs mer
Losmapimod, MedChemExpress
Produktkod. 16443928
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16443928 10 mg
16440287 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16443928 Leverantör Medchem Express Leverantörsnummer HY10402
MedChemExpress Losmapimod (GSK-AHAB) is a selective, potent, and orally active p38 MAPK inhibitor with pKis of 8.1 and 7.6 for p38α and p38β, respectively.
Losmapimod (GSK-AHAB) is a selective, potent, and orally active p38 MAPK inhibitor with pKis of 8.1 and 7.6 for p38α and p38β, respectively.
Specifikationer
Kemiskt namn eller material Losmapimod
CAS 585543-15-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C22H26FN3O2
Kvantitet 10 mg
Synonym GSK-AHAB GW856553X SB856553
Löslighetsinformation Ethanol : 33.33 mg/mL (86.92 mM; Need ultrasonic) ∣DMSO : 27.5 mg/mL (71.72 mM; Need ultrasonic)
LEDER CC1=C(C2=NC=C(C(NCC(C)(C)C)=O)C=C2)C=C(C(NC3CC3)=O)C=C1F
Molekylvikt (g/mol) 383.46
Formel vikt 383.46
Procent renhet 98.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation