Learn More
Thermo Scientific Chemicals Lovastatin, 98%
Use promo code "21615" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Applications
• It blocks product ion of mevalonate, a critical compound in the production of cholesterol and isoprenoids
• It can increase cellular resistance to anticancer agents such as doxorubicin • It can induce apoptosis in myeloma cells
• It can cause cell cycle arrest in G1 and G2/M phase
• It also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF
Specifications
Melting Point | 175.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: less soluble in ethanol, soluble in chloroform and acetone |
Formula Weight | 404.54 |
Percent Purity | 98% |
Chemical Name or Material | Lovastatin |
Safety and Handling
- Lovastatin
- Warning
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H317-May cause an allergic skin reaction.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only