Learn More
Lovibond™ Aqua Comparitor Color Discs: Aluminum
For testing water with the Aqua Comparator
Brand:  Lovibond™ 146200
Additional Details : Weight : 0.02900kg
Product Code. 11755448
  • 260.00 SEK / Each
Estimated Shipment: 17-10-2023
to see stock.
Add to basket
Description
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for aluminum using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™
0 to 0.3mg/L
Aluminum Color Disc