Learn More
Lovibond™ Aqua Comparitor Color Discs: Ozone
For testing water with the Aqua Comparator
Brand: Lovibond™ 146270
Additional Details : Weight : 0.03000kg
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for ozone using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0 to 0.7mg/L | |
Ozone Color Disc |