Learn More
Lovibond™ Aqua Comparitor Color Discs: Phosphate
For testing water with the Aqua Comparator
Brand: Lovibond™ 146240
Additional Details : Weight : 0.02800kg
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for phosphates using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0 to 4mg/L | |
Phosphate Color Disc |