Learn More
Lovibond™ MD600 Colorimeter Replacement Reagents: Chloride T1
For use with MD600 colorimeters and SpectroDirect spectrophotometer
Brand: Lovibond™ 515910BT
Additional Details : CAS Number : 7681-38-1 Weight : 0.09000kg
Specifications
White | |
Tablet | |
MC500, MD600 colorimeter and SP600 spectrophotometer | |
0.5 to 25mg/L | |
Silver Nitrate/Turbidity |
1.7 | |
Water quality testing | |
100 | |
20°C | |
Chloride T1 Tablet |
Safety and Handling
- Chloride T1
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium hydrogensulphate