Läs mer
3M™ Particulate Respirator, Series C100
Handla allt 3M Produkter
Filtertyp:
FFP1
FFP2
Med utandningsventil:
No
Yes
Förpackningsstorlek:
Paket med 10
Paket med 20
Beskrivning
Provides cost-effective, lightweight, comfortable, and effective protection against dust and mist.
- The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes
- Exhalation valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifikationer
Specifikationer
| Filtertyp | FFP2 |
| Material | Polypropylene |
| Produkttyp | General Purpose Particulate Respirator |
| Med utandningsventil | No |
| Modell | C102 |
| Sterilitet | Non-sterile |
| FDA godkänd kirurgisk mask | No |