Läs mer
Loxapine USP Related Compound A, Mikromol™
Produktkod. 30460679
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460679 4 x 25 mg
30049842 25 mg
30049429 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460679 Leverantör Mikromol Leverantörsnummer MM1530.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Loxapine USP Related Compound A, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Chloro-11-(4-methylpiperazin-1-yl)dibenzo[b,f][1,4]oxazepine
CAS 3454-96-4
Analyt- eller komponentnamn 3-Chloro-11-(4-methylpiperazin-1-yl)dibenzo[b,f][1,4]oxazepine
Frakt skick Room Temperature
Molekylformel C18 H18 Cl N3 O
Certifikat och efterlevnad ISO 17025
Synonym Dibenz[b,f][1,4]oxazepine, 3-chloro-11-(4-methyl-1-piperazinyl)-, Loxapine Related Compound A, Loxapine Related Compound A
InChI formel InChI=1S/C18H18ClN3O/c1-21-8-10-22(11-9-21)18-14-7-6-13(19)12-17(14)23-16-5-3-2-4-15(16)20-18/h2-7,12H,8-11H2,1H3
Lösningstyp Neat
LEDER CN1CCN(CC1)C2=Nc3ccccc3Oc4cc(Cl)ccc24
IUPAC-namn 9-chloro-6-(4-methylpiperazin-1-yl)benzo[b][1,4]benzoxazepine
Molekylvikt (g/mol) 327.81
Kvantitet 4 x 25 mg
Formel vikt 327.1138 g/mol