missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ LPA Monoclonal Antibody (4H1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517111
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA5-17111 targets LPA in indirect ELISA, ICC, IHC, IF and WB applications and shows reactivity with Human samples. MA5-17111 detects LPA which has a predicted molecular weight of approximately 501kDa.
The protein encoded by this gene is a serine proteinase that inhibits the activity of tissue-type plasminogen activator I. The encoded protein constitutes a substantial portion of lipoprotein(a) and is proteolytically cleaved, resulting in fragments that attach to atherosclerotic lesions and promote thrombogenesis. Elevated plasma levels of this protein are linked to atherosclerosis. Depending on the individual, the encoded protein contains 2-43 copies of kringle-type domains. The allele represented here contains 15 copies of the kringle-type repeats and corresponds to that found in the reference genome sequence.
Specifikationer
| LPA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P08519 | |
| LPA | |
| Purified recombinant fragment of human LPA (amino acids: 1823-2013) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4H1 | |
| Unconjugated | |
| LPA | |
| AK38; antiangiogenic AK38 protein; apo(a); APOA; apolipoprotein(a); lipoprotein(a); lipoprotein, Lp(a); LP; lp(a); LPA | |
| Mouse | |
| Protein G | |
| RUO | |
| 4018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering