missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRBA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27597-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the WDL-BEACH-WD (WBW) gene family. Its expression is induced in B cells and macrophages by bacterial lipopolysaccharides (LPS). The encoded protein associates with protein kinase A and may be involved in leading intracellular vesicles to activated receptor complexes, which aids in the secretion and/or membrane deposition of immune effector molecules. Defects in this gene are associated with the disorder common variable immunodeficiency-8 with autoimmunity. Two transcript variants encoding different isoforms have been found for this gene.Specifications
LRBA | |
Polyclonal | |
Unconjugated | |
LRBA | |
Beige like protein, BGL, CDC4 like protein, CDC4L, LAB300, LBA, LRBA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
987 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P50851 | |
LRBA | |
LRBA Fusion Protein Ag25596 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |