missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRBA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LRBA |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825725
|
Proteintech
27597-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815725
|
Proteintech
27597-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the WDL-BEACH-WD (WBW) gene family. Its expression is induced in B cells and macrophages by bacterial lipopolysaccharides (LPS). The encoded protein associates with protein kinase A and may be involved in leading intracellular vesicles to activated receptor complexes, which aids in the secretion and/or membrane deposition of immune effector molecules. Defects in this gene are associated with the disorder common variable immunodeficiency-8 with autoimmunity. Two transcript variants encoding different isoforms have been found for this gene.Specifications
LRBA | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P50851 | |
987 | |
LRBA Fusion Protein Ag25596 | |
Primary | |
-20°C | |
LRBA |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Beige like protein, BGL, CDC4 like protein, CDC4L, LAB300, LBA, LRBA | |
LRBA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |