missing translation for 'onlineSavingsMsg'
Learn More

LRP1 Recombinant Human Protein

LRP1 Recombinant Human Protein

Brand:  Invitrogen 11665HCCH25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238815

  • 1464.00 SEK / 25µg


Estimated Shipment date
from Supplier 05-06-2019
Add to basket

Description

Description

Low Density Lipoprotein Receptor-Related Protein 1 (LRP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human (Arg 786-Leu 1165) LRP1 (NP_002323.2) corresponding to the cluster II LDLR domain fused to a peptide (ALEVLFQ) at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein assays and analysis
Lyophilized
42kDa
PAGE
5 x 5μg
For research use only. Not for use in diagnostic procedures
A2MR, APOER, APR, CD91, FLJ16451, IGFBP3R, LRP, MGC88725,TGFBR5
5 Vials of protein in suspension buffer
Assay standard
Mammalian
Human
Lyophilized
4035
LRP1 Recombinant Human Protein
≥90% by SDS-PAGE
–20°C
5 Pack(s)
LRP1 Recombinant Human Protein
LRP1
CD proteins
LRP1 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.