missing translation for 'onlineSavingsMsg'
Learn More

LRP1 Recombinant Human Protein

LRP1 Recombinant Human Protein

Brand:  Invitrogen 11665HCCH5

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258815

  • 681.36 SEK / 5µg


Estimated Shipment date
from Supplier 04-02-2019
Add to basket

Description

Description

Low Density Lipoprotein Receptor-Related Protein 1 (LRP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human (Arg 786-Leu 1165) LRP1 (NP_002323.2) corresponding to the cluster II LDLR domain fused to a peptide (ALEVLFQ) at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein assays and analysis
Lyophilized
42kDa
PAGE
5μg
For research use only. Not for use in diagnostic procedures
A2MR, APOER, APR, CD91, FLJ16451, IGFBP3R, LRP, MGC88725,TGFBR5
1 Vial of protein in suspension buffer
Assay standard
Mammalian
Human
Lyophilized
4035
LRP1 Recombinant Human Protein
≥90% by SDS-PAGE
–20°C
1 Each
LRP1 Recombinant Human Protein
LRP1
CD proteins
LRP1 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.