Learn More
LRRC33 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556788
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PSCVDFRNMA SLRSLSLEGC GLGALPDCPF QGTSLTYLDL SSNWGVLNGS LAPLQDVAPM LQVLSLRNMG LHSSFMALDF SGFGNLRDLD LSGNCLTTFP RFGGSLALET LDLRRNSLTA LPQKAVSEQL SRGLRTIYLS QN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
Specifications
LRRC33 | |
Polyclonal | |
Unconjugated | |
NRROS | |
GARPL1, UNQ3030, ELLP3030, leucine-rich repeat-containing protein 33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375387 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YC3 | |
NRROS | |
Recombit protein corresponding to Human LRRC33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only