missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC8D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LRRC8D |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838683
|
Proteintech
11537-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828683
|
Proteintech
11537-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This LRRC8D antibody does not cross-react with LRRC8A.
Component of the volume-regulated anion channel (VRAC, also named VSOAC channel), an anion channel required to maintain a constant cell volume in response to extracellular or intracellular osmotic changes. The VRAC channel conducts iodide better than chloride and may also conduct organic osmolytes like taurine. It is unclear whether LRRC8D constitutes a pore-forming subunit or whether it is closely associated with the pore.Specifications
LRRC8D | |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q5U308, Q7L1W4, Q8BGR2 | |
231549, 305131, 55144 | |
LRRC8D Fusion Protein Ag2138 | |
Primary | |
-20°C | |
LRRC8D |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
LRRC5, LRRC8D | |
Lrrc8d | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |