missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ LRRK1 Polyclonal Antibody

Beskrivning
Antibody detects endogenous levels of total LRRK1.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Specifikationer
Specifikationer
| Antigen | LRRK1 |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | Lrrk1 |
| Genaccessionsnr. | Q38SD2, Q3UHC2 |
| Gene Alias | AW319595; C230002E15Rik; D130026O16Rik; Kiaa1790; leucine rich repeat kinase 1; leucine-rich repeat kinase 1; leucine-rich repeat serine/threonine-protein kinase 1; LRRK1; mKIAA1790; RIPK6; Roco1 |
| Gensymboler | Lrrk1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?