Läs mer
LRRK2 inhibitor 1, MedChemExpress
Produktkod. 16475346
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16475346 100 mg -
16467197 5 mg -
16450787 50 mg 50mg
16424478 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16475346 Leverantör Medchem Express Leverantörsnummer HY111493
MedChemExpress LRRK2 inhibitor 1 is a potent, selective and oral LRRK2 inhibitor with an pIC50 of 6.8.
LRRK2 inhibitor 1 is a potent, selective and oral LRRK2 inhibitor with an pIC50 of 6.8.
Specifikationer
Kemiskt namn eller material LRRK2 inhibitor 1
CAS 1802525-61-6
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C20H23N5O4
Kvantitet 100 mg
Löslighetsinformation DMSO : 62.5 mg/mL (157.26 mM; Need ultrasonic)
LEDER CCOC1=NC(NC2=C(OC)C=C(C(N3CCOCC3)=O)C=C2)=NC4=C1C=CN4
Molekylvikt (g/mol) 397.43
Formel vikt 397.43
Procent renhet 99.72%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation