missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRTM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14288-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM1 is a maternally suppressed gene that is associated paternally with handedness and schizophrenia.Specifications
LRRTM1 | |
Polyclonal | |
Unconjugated | |
LRRTM1 | |
LRRTM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
347730, 679668, 74342 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D4A6D8, Q86UE6, Q8K377 | |
LRRTM1 | |
LRRTM1 Fusion Protein Ag5602 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |