Learn More
Corning™ LSE™ Shaking Incubator Accessory, Plastic Clamps
Product Code. 16813440 Shop All Corning Products
Change view
Click to view available options
Holds:
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
16813440 1000 mL Each
16803440 125 mL -
16279601 250 mL Each
16289601 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16813440 Supplier Corning™ Supplier No. 480152
  • ONLINE EXCLUSIVE PRICE
    683.39 SEK
    634.53 SEK / Paket med 1
    Save 48.86 SEK
    7% Av
Beräknad leverans: 08-04-2027
For use with LSE Shaking Incubator
Constructed of polypropylene.
TRUSTED_SUSTAINABILITY
Specifications
Holds 1000 mL
Product Type Clamp
For Use With (Equipment) LSE Benchtop Shaking Incubator