Learn More
LSM1 Antibody, PerCP, R&D Systems™
Product Code. 18606698 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18606698 0.1mL
1 options
Product Code. 18606698 Supplier Novus Biologicals Supplier No. NBP300249PCP
 Restricted Item
Beräknad leverans: 10-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
LSM1 Polyclonal specifically detects LSM1 in Human samples. It is validated for ELISA/Immunohistochemistry/Immunohistochemistry (Paraffin)/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Product Type Antibody
Product Line Novus