missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTBR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | LTBR |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839854
|
Proteintech
20331-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829854
|
Proteintech
20331-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin alpha and lymphtoxin beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.Specifications
LTBR | |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P36941, P50284 | |
17000, 4055 | |
LTBR Fusion Protein Ag14065 | |
Primary | |
-20°C | |
LTBR |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D12S370, LT BETA R, LTBR, Lymphotoxin beta receptor, TNF R III, TNF RIII, TNFCR, TNFR III, TNFR RP, TNFR2 RP, TNFR3, TNFRSF3 | |
Ltbr | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |