Läs mer
LTURM34, MedChemExpress
Produktkod. 16489956
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16489956 50 mg
16473488 10 mg
16426217 5 mg
16404666 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16489956 Leverantör Medchem Express Leverantörsnummer HY101667
MedChemExpress LTURM34 is a specific DNA-PK inhibitor (IC50=34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines.
LTURM34 is a specific DNA-PK inhibitor (IC50=34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines.
Specifikationer
Kemiskt namn eller material LTURM34
CAS 1879887-96-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C24H18N2O3S
Kvantitet 50 mg
Löslighetsinformation DMSO : 65 mg/mL (156.82 mM; Need ultrasonic)
LEDER O=C1C2=CC=CC(C3=CC=CC4=C3SC5=C4C=CC=C5)=C2OC(N6CCOCC6)=N1
Molekylvikt (g/mol) 414.48
Formel vikt 414.48
Procent renhet 99.89%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease