Läs mer
Lucanthone Hydrochloride, LoGiCal
Produktkod. 30457630
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30457630 10 mg
1 options
Produktkod. 30457630 Leverantör LoGiCal Leverantörsnummer LGCFOR1641.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lucanthone Hydrochloride
CAS 548-57-2
Analyt- eller komponentnamn Lucanthone Hydrochloride
Frakt skick Room Temperature
Molekylformel C20 H24 N2 O S . Cl H
Certifikat och efterlevnad ISO 9001
Synonym 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (9CI), Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, monohydrochloride (6CI,8CI), 1-[(2-Diethylaminoethyl)amino]-4-methylthioxanthen-9-one hydrochloride, 3735 R. P., 79T61, BW 57-233, Lucanthone hydrochloride, Lucanthone monohydrochloride, Miracil D, Miracol, Ms. 752, NSC 14574, Nilodin, RP 3735, Scapuren, Tixantone, 9H-Thioxanthen-9-one, 1-[[2-(diethylamino)ethyl]amino]-4-methyl-, hydrochloride (1:1), Lucanthone Hydrochloride
InChI formel InChI=1S/C20H24N2OS.ClH/c1-4-22(5-2)13-12-21-16-11-10-14(3)20-18(16)19(23)15-8-6-7-9-17(15)24-20;/h6-11,21H,4-5,12-13H2,1-3H3;1H
Lösningstyp Neat
LEDER Cl.CCN(CC)CCNc1ccc(C)c2Sc3ccccc3C(=O)c12
IUPAC-namn 1-[2-(diethylamino)ethylamino]-4-methylthioxanthen-9-one;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 376.94
Kvantitet 10 mg
Formel vikt 376.1376 g/mol