Läs mer
Lucidenic Acid B, TRC
Produktkod. 30479657
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30479657 5 mg
1 options
Produktkod. 30479657 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4737205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lucidenic Acid B
CAS 95311-95-8
Molekylformel C27H38O7
Kvantitet 5 mg
Synonym (+)-Lucidenic Acid B, (5α,7β,12β)-7,12-Dihydroxy-4,4,14-trimethyl-3,11,15-trioxochol-8-en-24-oic Acid
InChI formel InChI=1S/C27H38O7/c1-13(7-8-19(31)32)14-11-18(30)27(6)20-15(28)12-16-24(2,3)17(29)9-10-25(16,4)21(20)22(33)23(34)26(14,27)5/h13-16,23,28,34H,7-12H2,1-6H3,(H,31,32)/t13-,14-,15+,16+,23-,25+,26+,27+/m1/s1
LEDER C[C@H](CCC(=O)O)[C@H]1CC(=O)[C@@]2(C)C3=C(C(=O)[C@@H](O)[C@]12C)[C@@]4(C)CCC(=O)C(C)(C)[C@@H]4C[C@@H]3O
Molekylvikt (g/mol) 474.59