missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luciferase Rabbit anti-Renilla, Tag, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Luciferase |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825945
|
Proteintech
27986-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815945
|
Proteintech
27986-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects the Luciferase protein of Photinus pyralis (Common eastern firefly) (Lampyris pyralis).
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.Specifications
Luciferase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Renilla | |
Luciferase, Luciferin 4-monooxygenase | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Luciferase Fusion Protein Ag27633 | |
Primary | |
-20°C |