Läs mer
Lumiflavin 5-Oxide, TRC
Produktkod. 30012100
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012100 25 mg
30008659 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012100 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47395025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36995-93-4
Molekylformel C13H12N4O3
Molekylvikt (g/mol) 272.26
Synonym 7,8,10-Trimethyl-benzo[g]pteridine-2,4(3H,10H)-dione 5-Oxide
LEDER CC1=CC2=C(C=C1C)[N+](=C3C(=O)NC(=O)N=C3N2C)[O-]
Specifikationer
Kemiskt namn eller material Lumiflavin 5-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C13H12N4O3/c1-6-4-8-9(5-7(6)2)17(20)10-11(16(8)3)14-13(19)15-12(10)18/h4-5H,1-3H3,(H,15,18,19)
Formel vikt 272.0909