Läs mer
Lumiracoxib Acyl-Beta-D-glucuronide, TRC
Produktkod. 30468385
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30468385 1 mg
1 options
Produktkod. 30468385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4747101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lumiracoxib Acyl-beta-D-glucuronide
CAS 697287-17-5
Molekylformel C21 H21 Cl F N O8
Kvantitet 1 mg
Synonym Lumiracoxib Acyl-B-D-Glucuronide
InChI formel InChI=1S/C21H21ClFNO8/c1-9-5-6-13(24-15-11(22)3-2-4-12(15)23)10(7-9)8-14(25)31-21-18(28)16(26)17(27)19(32-21)20(29)30/h2-7,16-19,21,24,26-28H,8H2,1H3,(H,29,30)/t16-,17-,18+,19-,21+/m0/s1
LEDER Cc1ccc(Nc2c(F)cccc2Cl)c(CC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)c1
IUPAC-namn (2S,3S,4S,5R,6S)-6-[2-[2-(2-chloro-6-fluoroanilino)-5-methylphenyl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 469.84
Formel vikt 469.094