Läs mer
Lumisterol, TRC
Produktkod. 30476628
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476628 10 mg
30476988 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30476628 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47480010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Lumisterol
CAS 474-69-1
Molekylformel C28H44O
Kvantitet 10 mg
Synonym Ergosta-5,7,22-trien-3-ol, (3β,9β,10α,22E)-, 9β,10α-Ergosta-5,7,22-trien-3-ol (7CI), 9β,10α-Ergosta-5,7,22-trien-3β-ol (8CI), Lumisterol (6CI), (3β,9β,10α,22E)-Ergosta-5,7,22-trien-3-ol, 9β,10α-Ergosterol, Lumisterol2, (22E)-9β,10α-Ergosta-5,7,22-trien-3β-ol
InChI formel InChI=1S/C28H44O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,18-20,22,24-26,29H,11-17H2,1-6H3/b8-7+/t19-,20+,22-,24+,25-,26+,27+,28+/m0/s1
LEDER CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@@]4(C)[C@@H]3CC[C@]12C
Molekylvikt (g/mol) 396.65
Formel vikt 396.3392