missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LW 6
GREENER_CHOICE

Product Code. 18736206
Click to view available options
Kvantitet:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18736206

Brand: Tocris Bioscience™ 6322/50

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Malate dehydrogenase-2 (MDH2) inhibitor; also inhibits HIF-1α

LW 6 is a malate dehydrogenase-2 (MDH2) inhibitor (IC50 = 6.3 μM). Also inhibits HIF-1α. Prevents accumulation of HIF-1α in Hep3B cells under hypoxic conditions (IC50 = 2.6 μM) with resulting decrease in expression of HIF-1α target genes VEGF and EPO. Inhibits proliferation of activated human T cells ex vivo.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methyl 4-hydroxy-3-[[2-(4-tricyclo[3.3.1.13,7 ]dec-1-ylphenoxy)acetyl]amino]benzoate
CAS 934593-90-5
Kvantitet 50 mg
Mål Dehydrogenase Inhibitors
Molekylformel C26H29NO5
Renhet 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.