Läs mer
LX-1031, MedChemExpress
Produktkod. 16493346
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493346 25 mg
16486086 100 mg
16485508 10 mg
16441687 50 mg
16428227 5 mg
16400196 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16493346 Leverantör Medchem Express Leverantörsnummer HY13041
MedChemExpress LX-1031 is a potent, orally available tryptophan 5-hydroxylase (TPH) inhibitor that reduces serotonin (5-HT) synthesis peripherally.
LX-1031 is a potent, orally available tryptophan 5-hydroxylase (TPH) inhibitor that reduces serotonin (5-HT) synthesis peripherally.
Specifikationer
Kemiskt namn eller material LX-1031
CAS 945976-76-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C28H25F3N4O4
Kvantitet 25 mg
Löslighetsinformation DMSO : ≥ 34 mg/mL (63.14 mM)
LEDER NC1=NC(C2=CC=C(C=C2)C[C@@H](C(O)=O)N)=CC(O[C@H](C3=CC=C(C4=CC(OC)=CC=C4)C=C3)C(F)(F)F)=N1
Molekylvikt (g/mol) 538.52
Formel vikt 538.52
Procent renhet 98.32%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation