Learn More
R&D Systems™ LXR beta/NR1H2 Antibody Blocking Peptide
Product Code. 18265017 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Product Code. Quantity Unit Size
18265017 0.05 mg 0.05mg
1 options
Product Code. 18265017 Supplier R&D Systems™ Supplier No. NBP176256PEP
 Restricted Item
Beräknad leverans: 02-12-2026
A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A blocking peptide containing 14 amino acids near the amino terminus of human LXR-B. Source: Synthetic (Accession #: P55055) The LXR beta/NR1H2 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 7376
Species Human
Product Line Novus
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76256. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias Liver x receptor beta, LXR-b, NER-I, NERliver x receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta
Gene Symbol NR1H2
Product Type LXR beta/NR1H2
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 14 amino acids near the amino terminus of human LXR-B