Läs mer
R&D Systems™ LY 2087101
Produktkod. 18075542
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18075542 10 mg
18044892 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18075542 Leverantör R&D Systems™ Leverantörsnummer 4141/10
 Begränsad artikel
Beräknad leverans: 27-11-2026
A Tocris product, part of R&D Systems. Potentiator of α7 nAChRs
Allosteric potentiator of α7, α4β2 and α4β4 nAChRs; displays selectivity against α3β4 nAChRs. Thought to potentiate agonist-evoked α7 responses by binding within the nAChR transmembrane region.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 913186-74-0
Molekylformel C15H11FN2OS2
Specifikationer
Kemiskt namn eller material LY 2087101
Kvantitet 10 mg
Produktlinje Tocris
Formel vikt 318.39
Renhet >98%
Molekylvikt (g/mol) 318.384