Läs mer
Tocris Bioscience™ LY 2109761
Beskrivning
LY 2109761 is a selective dual TGF-β receptor type I and II (TβRI/II) inhibitor (Ki values are 38 and 300 nM in a cell-free assay, respectively); it inhibits the kinase activity of both TGF-βRI and TGF-βRII. In vitro, it significantly inhibits the growth, motility, and invasiveness of L3.6pl/GLT pancreatic cancer cells and completely suppresses TGF-β-induced Smad2 phosphorylation. In vivo, LY 2109761, in combination with Gemcitabine, significantly reduces tumor volume and spontaneous abdominal metastases in a murine model of metastatic pancreatic cancer. LY 2109761 is orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[5,6-Dihydro-2-(2-pyridinyl)-4 H-pyrrolo[1,2-b]pyrazol-3-yl]-7-[2-(4-morpholinyl)ethoxy]quinoline |
| CAS | 700874-71-1 |
| Kvantitet | 50 mg |
| Mål | TGF-beta Receptor Inhibitors |
| Molekylformel | C26H27N5O2 |
| Renhet | 0.98 |