Läs mer
LY-338979, TRC
Produktkod. 30475102
Change view
Kvantitet:
1 mg
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30475102 1 mg -
30474610 25 mg -
30469101 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475102 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL4866501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 193281-00-4
Molekylformel C20 H21 N5 O7
Molekylvikt (g/mol) 443.41
Synonym N-[4-[2-(2-Amino-4,5,6,7-tetrahydro-4,6-dioxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid, LY 338979
IUPAC-namn (2S)-2-[[4-[2-(2-amino-4,6-dioxo-5,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid
LEDER NC1=NC2=C(C(CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)N2)C(=O)N1
Specifikationer
Kemiskt namn eller material LY-338979
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C20H21N5O7/c21-20-24-15-14(18(30)25-20)11(17(29)23-15)6-3-9-1-4-10(5-2-9)16(28)22-12(19(31)32)7-8-13(26)27/h1-2,4-5,11-12H,3,6-8H2,(H,22,28)(H,26,27)(H,31,32)(H4,21,23,24,25,29,30)/t11?,12-/m0/s1
Formel vikt 443.1441
Procent renhet >95%