Läs mer
Tocris Bioscience™ LY 344864 hydrochloride
GREENER_CHOICE
Produktkod. 18035352
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18035352 10 mg
18014382 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18035352 Leverantör Tocris Bioscience™ Leverantörsnummer 2451/10
Potent and selective 5-HT1F agonist
Potent, selective 5-HT1F receptor agonist (EC50 = 3 nM). Displays > 80-fold selectivity over other 5-HT receptors (Ki values are 0.006, 0.53, 0.55, 0.56, 1.42, 1.70, 3.50, 3.94 and 4.85 μM for 5-HT1F, 5-HT1A, 5-HT1B, 5-HT1D, 5-HT1E, 5-HT2B, 5-HT2C, 5-HT2A and 5-HT7 receptors respectively). Inhibits neurogenic dural inflammation in vivo following i.v. and oral administration.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217756-94-9
Molekylformel C21H23ClFN3O
Specifikationer
Kemiskt namn eller material LY 344864 hydrochloride
Kvantitet 10 mg
Formel vikt 387.88
Renhet >99%
Molekylvikt (g/mol) 387.883