Läs mer
LY 344864, MedChemExpress
Produktkod. 16492007
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492007 50 mg
16485858 10 mg
16418517 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16492007 Leverantör Medchem Express Leverantörsnummer HY13788
MedChemExpress LY 344864 is a selective receptor agonist with an affinity of 6 nM (Ki) at the recently cloned 5-HT1F receptor.
LY 344864 is a selective receptor agonist with an affinity of 6 nM (Ki) at the recently cloned 5-HT1F receptor.
Specifikationer
Kemiskt namn eller material LY 344864
CAS 186544-26-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C21H22FN3O
Kvantitet 50 mg
Löslighetsinformation DMSO : 100 mg/mL (284.56 mM; Need ultrasonic)
LEDER O=C(NC1=CC2=C(NC3=C2C[C@H](N(C)C)CC3)C=C1)C4=CC=C(F)C=C4
Molekylvikt (g/mol) 351.42
Formel vikt 351.42
Procent renhet 98.93%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation