Läs mer
LY 368962 Dimethyl Ester, TRC
Produktkod. 30475361
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475361 100 mg
1 options
Produktkod. 30475361 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL486705100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[4-[3-(2,6-Diamino-1,4-dihydro-4-oxo-5-pyrimidinyl)-3-oxopropyl]benzoyl]-L-glutamic Acid Dimethyl Ester
CAS 1798887-66-7
Molekylformel C21H25N5O7
Kvantitet 100 mg
Synonym N-[4-[3-(2,6-Diamino-1,4-dihydro-4-oxo-5-pyrimidinyl)-3-oxopropyl]benzoyl]-L-glutamic Acid Dimethyl Ester
InChI formel InChI=1S/C21H25N5O7/c1-32-15(28)10-8-13(20(31)33-2)24-18(29)12-6-3-11(4-7-12)5-9-14(27)16-17(22)25-21(23)26-19(16)30/h3-4,6-7,13H,5,8-10H2,1-2H3,(H,24,29)(H5,22,23,25,26,30)/t13-/m0/s1
LEDER O=C(OC)CC[C@@H](C(OC)=O)NC(C1=CC=C(CCC(C2=C(N)NC(N)=NC2=O)=O)C=C1)=O
IUPAC-namn dimethyl (4-(3-(2,6-diamino-4-oxo-1,4-dihydropyrimidin-5-yl)-3-oxopropyl)benzoyl)-L-glutamate
Molekylvikt (g/mol) 459.45
Formel vikt 459.18