Läs mer
Tocris Bioscience™ LY 487379 hydrochloride
GREENER_CHOICE
Produktkod. 18034252
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18034252 10 mg
18094432 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18034252 Leverantör Tocris Bioscience™ Leverantörsnummer 3283/10
Selective positive allosteric modulator of mGlu2 receptors
Positive allosteric modulator selective for mGlu2 receptors. Potentiates glutamate-stimulated [35S]GTPγS binding (EC50 values are 1.7 and > 10 μM for mGlu2 and mGlu3 receptors respectively). Devoid of any activity at mGlu5 and mGlu7 receptors. Promotes cognitive flexibility in a rat model.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 353229-59-1
Molekylformel C21H20ClF3N2O4S
Specifikationer
Kemiskt namn eller material LY 487379 hydrochloride
Kvantitet 10 mg
Formel vikt 488.91
Renhet >99%
Molekylvikt (g/mol) 488.906