missing translation for 'onlineSavingsMsg'
Learn More

Ly-49H Mouse anti-Mouse, PE-Cyanine7, Clone: 3D10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5886-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15568586

  • 718.00 SEK / 25µg


Estimated Shipment
11-01-2021
Add to basket

Description

Description

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Ly49H is a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins.
Specifications

Specifications

Ly-49H
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q60682
Klra8
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16639
Flow Cytometry
3D10
PE-Cyanine7
Klra8
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
SDS
Documents

For Research Use Only.