Läs mer
LY3502970, MedChemExpress
Produktkod. 16454598
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16454598 10 mg
16450897 50 mg
16445446 100 mg
16413156 25 mg
16407317 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16454598 Leverantör Medchem Express Leverantörsnummer HY112185
MedChemExpress LY3502970 (GLP-1 receptor agonist 1) is a GLP-1 receptor agonist extracted from patent WO2018056453A1, Compound 67.
LY3502970 (GLP-1 receptor agonist 1) is a GLP-1 receptor agonist extracted from patent WO2018056453A1, Compound 67.
Specifikationer
Kemiskt namn eller material LY3502970
CAS 2212020-52-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C48H48F2N10O5
Kvantitet 10 mg
Synonym GLP-1 receptor agonist 1
Löslighetsinformation DMSO : 125 mg/mL (141.57 mM; Need ultrasonic)
LEDER C[C@@H]1[C@](N2C3=CC=C([C@]4([H])CC(C)(OCC4)C)C=C3C=C2C(N5[C@H](C6=C(N7C(N(C8=CC=C(N(N=C9)C)C9=C8F)C=C7)=O)N(C%10=CC(C)=C(C(C)=C%10)F)N=C6CC5)C)=O)(C(N%11)=NOC%11=O)C1
Molekylvikt (g/mol) 882.96
Formel vikt 882.96
Procent renhet 98.02%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Metabolism-sugar/lipid metabolism