Läs mer
LY450108, MedChemExpress
Produktkod. 16437017
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16437017 5 mg
16430627 50 mg
16424298 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16437017 Leverantör Medchem Express Leverantörsnummer HY10935
MedChemExpress LY450108 is a potent AMPA receptor potentiator. LY450108 has the potential for depression and Parkinson's disease research.
LY450108 is a potent AMPA receptor potentiator. LY450108 has the potential for depression and Parkinson's disease research.
Specifikationer
Kemiskt namn eller material LY450108
CAS 376594-67-1
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C19H22F2N2O3S
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 50 mg/mL (126.12 mM) ∣H2O : 1 mg/mL (2.52 mM; Need ultrasonic and warming)
LEDER FC1=CC(F)=CC(C(NC2=CC=C([C@@H](C)CNS(C(C)C)(=O)=O)C=C2)=O)=C1
Molekylvikt (g/mol) 396.45
Formel vikt 396.45
Procent renhet 97.7%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation