missing translation for 'onlineSavingsMsg'
Learn More

LY6G6F Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25872-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824435

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May play a role in the downstream signal transduction pathways involving GRB2 and GRB7.
Specifications

Specifications

LY6G6F
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5SQ64
C6orf21, G6f, LY6G6D, LY6G6F, NG32
Rabbit
IgG
20 μL
-20°C
Primary
259215
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
LY6G6F
Liquid
LY6G6F
LY6G6F Fusion Protein Ag22467
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers