missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LY6H (Human) Recombinant Protein

Human LY6H full-length ORF ( AAH28894, 26 a.a. - 140 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004062-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177481

  • 3343.96 SEK / 10µg

Estimated Shipment: 14-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: LWCQDCTLTTNSSHCTPKQCQPSDTVCASVRITDPSSSRKDHSVNKMCASSCDFVKRHFFSDYLMGFINSGILKVDVDCCEKDLCNGAAGAGHSPWALAGGLLLSLGPALLWAGP
Specifications

Specifications

AAH28894
lymphocyte antigen 6 complex, locus H
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LY6H
GST
4062
Wheat germ expression system
10 μg
NMLY6
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates