missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY96/MD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11784-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS. The specificity of this response is determined by the species of MD-2; e. g., human MD-2 transfected into mouse cells can cause mouse TLR4 to react to LPS analogs that are normally antagonistic to human but not mouse TLR4.Specifications
LY96/MD2 | |
Polyclonal | |
Unconjugated | |
LY96 | |
ESOP 1, ESOP1, Ly 96, LY96, lymphocyte antigen 96, MD 2, MD2, Protein MD 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23643 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y6Y9 | |
LY96 | |
LY96/MD2 Fusion Protein Ag2447 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |