Läs mer
Lymecycline (85%), TRC
Produktkod. 30009663
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009663 100 mg
30006849 5 mg
30010188 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009663 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL487800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 992-21-2
Molekylformel C29 H38 N4 O10
Molekylvikt (g/mol) 602.63
Synonym L-Lysine, N6-[[[[(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenyl]carbonyl]amino]methyl]-, L-Lysine, N6-[[[[4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenyl]carbonyl]amino]methyl]-, [4S-(4α,4aα,5aα,6β,12aα)]-, Lysine, N6-[[4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamido]methyl]- (7CI), Lysine, N6-[[4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamido]methyl]-, L- (8CI), (+)-N-(5-Amino-5-carboxypentylaminomethyl)-4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxonaphthacene-2-carboxamide, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, lysinemethylene deriv., Armyl, Ciclisin, Ciclolysal, Ciclolysine, Infaciclina, Lisinbiotic, Lisinciclina, Lymecycline, Mucomycin, N-Lysinomethyltetracycline, N2-[[(+)-5-Amino-5-carboxypentylamino]methyl]tetracycline, N6-[[4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamido]methyl]lysine, Tertamyl, Tetracycline-L-methylenelysine, Tetracyclinemethylenelysine, Tetralisal, Tetralysal, Tetramyl, Vebicyclysal
IUPAC-namn (2S)-6-[[[(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carbonyl]amino]methylamino]-2-aminohexanoic acid
LEDER CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)NCNCCCC[C@H](N)C(=O)O)C(=O)c4c(O)cccc4[C@@]3(C)O
Specifikationer
Kemiskt namn eller material Lymecycline
Kvantitet 100 mg
InChI formel InChI=1S/C29H38N4O10/c1-28(42)13-7-6-9-17(34)18(13)22(35)19-14(28)11-15-21(33(2)3)23(36)20(25(38)29(15,43)24(19)37)26(39)32-12-31-10-5-4-8-16(30)27(40)41/h6-7,9,14-16,21,31,34,36-37,42-43H,4-5,8,10-12,30H2,1-3H3,(H,32,39)(H,40,41)/t14-,15-,16-,21-,28+,29-/m0/s1
Formel vikt 602.2588