Learn More
Alfa Aesar™ [Lys&superscript_1;, Pro&superscript_2;,&superscript_5;, Arg&superscript_3;,&superscript_4;, Tyr&superscript_6;] Vasoactive Intestinal Peptide, Human, Porcine, Rat, Ovine
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J66201.|D
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15465805
  • 1118.41 SEK / 0.50mg
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Disclaimers:
For Research Use Only
Specifications
Specifications
3467.10
Human, Porcine, Rat, Ovine
Solid
RUO
[Lys1, Pro2,5,Arg3,4,Tyr6] VIP, human, porcine, rat, ovine
HPLC
0.5mg
Lys-Pro-Arg-Arg-Pro-Tyr-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
-30° to -10°C
[Lys1, Pro2,5, Arg3,4, Tyr6] Vasoactive Intestinal Peptide, human, porcine, rat, ovine